missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 604 Surface Protection Paper

Product Code. 11725375 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W):
50 m x 46 cm
50 m x 6 cm
520 x 420 mm
570 x 460 mm
600 x 500 mm
Packaging Type:
Roll
Sheet
Unit Size:
Each
Pack of 100
This item is not returnable. View return policy

Product Code. 11725375

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Designed to protect laboratory surfaces against hazardous spills. Fisherbrand™ Grade 604 Surface Protection Paper guards against damage from acids, toxic, corrosive and radioactive fluids and impacts.

  • Available in sheet or roll form
  • Polyethylene backed 125g/m2 absorbent paper quickly soaks up spills and prevents flow through to work surface

Specifications

Product Type Surface Protection Paper
Material Grade 604 Paper
Length (Metric) 600 mm
Width (Metric) 500 mm
Dimensions (L x W) 600 x 500 mm
Backing Material Polyethylene
Color White
Thickness 0.25mm
Quantity 100 sheets
Packaging Type Sheet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.