Learn More
Fisherbrand™ Grade 508 Chromatography Paper
Product Code. 11724166 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
200 mm
570 mm
Width (Metric):
200 mm
460 mm
Unit Size:
Pack of 100
Pack of 50
Product Code. 11724166
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Suitable for electrophoresis and chromatographic works. Fisherbrand™ Grade 508 Chromatography Paper is used in separation and identification of samples unknown composition in research laboratories.
Applications:
  • Used in separation and identification of additives in food
  • Helps in determination of malic acid content in wines caused by fermentation
Specifications
Film Thickness 0.35 mm
For Use With (Application) Suitable for Electrophoresis and Chromatographic Works
Format Sheet
Grade 508
Material Paper
Width (Metric) 200 mm
Flow Rate 115 mm/30 min.
Length (Metric) 200 mm