missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPBAR-A, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
GPBA receptor agonist. Upregulates GLP-1 secretion from intestinal cultures. Increases MQAE-fluorescence suggesting a decrease in intracellular chloride concentration in gallbladder epithelial cells.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 484.37 |
Percent Purity | >98% |
Chemical Name or Material | GPBAR-A |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'