Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
Click to view available options
Capacity:
101 kg
101 kg/61 kg
12 kg
21 kg
31 kg
31 kg/6.1 kg
61 kg
Readability:
0.1 g
1 g
1 g/0.1 g (smart range)
10 g/1 g (smart range)
Unit Size:
Each
Description
- EC Type approved
- Swing arm design or separate display and platform design
- IP65 protected
- Durable aluminum alloy & stainless steel construction
- Easy calibration with internal calibration mass
- SHS response speed of only 1.5 seconds
- GLP, GMP and ISO compliant
- RS-232C interface
- Comparator output and analog output (optional)
- Large Vacuum Fluorescent display
- Data memory function
- Counting function with ACAI (Automatic Counting Accuracy Improvement)
- Smart range function (GP-32K/102K/32KS)
- Comparator function
- Underhook weighing for thermal and magnetic samples as well as for density calculation
- Animal weighing function
Specifications
Specifications
| Capacity | 101 kg |
| Display Type | VFD |
| Readability | 1 g |
| Stabilization Time | 1.5 seconds |
| Type | Industrial Precision Balance |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering