Learn More
Ismatec™ Gore Style 100CR Tubing
Click to view available options
Diameter (Metric) Inner:
2.8 mm
Unit Size:
Each
Description
Order Info
DiameterMetricInside
Specifications
Specifications
| Diameter (Metric) Inner | 2.8 mm |
| Material | Gore 100CR |
| Diameter (Metric) Outer | 4.4 mm |
| Overall Length (Metric) | 30.5 cm |
| Packaging Quantity | 1 Each |
| Color | White |
| Product Type | Tubing |