Learn More
SCHOTT AG Lighting and Imaging™ Gooseneck Light Guide for KL300 LED
Fiber optic flex and stay light guide in a black lacquered semi-rigid gooseneck tubing.
Brand:  SCHOTT AG Lighting and Imaging™ 170202
 View more versions of this product
Product Code. 11842442
  • 273.00€ / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Light guide with eco-friendly Puravis fiber optics.
  • Precise positioning offers complete control of light placement.
  • Gooseneck branches positioned in opposite directions create near shadow-free illumination.
  • Re-adjust with ease.
  • For use with Cold light source KL300LED.
  • Diameter: End tip: 6 mm, Lightsource adapter: 9 mm, Active Fiber optic bundle: 4.5 mm, Outer Jacketing: 6.5 mm
Specifications
Specifications
500 mm