missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Goat Serum

Suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.

45.50€

Specifications

Color Reddish brown
Content And Storage Store at -20°C
Form Liquid
Sterilization Method 0.2 μm filtration
Source Whole blood collected from healthy adult animals
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11475055
View Documents
MP Biomedicals™
092939149
100 mL
45.50€
100mL
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
Description

Description

This serum is produced from whole blood collected from healthy adult animals.

Specifications

Specifications

Reddish brown
Liquid
Whole blood collected from healthy adult animals
7 to 8
Store at -20°C
0.2 μm filtration
Cell Culture
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.