Learn More
IgG Goat anti-Rabbit, Brilliant Violet 421, Polyclonal, BD
Goat Polyclonal Antibody
Brand: BD Biosciences 565014
Description
BD Horizon™ BV421 Goat Anti-Rabbit IgG is intended to be a second-step reagent for immunofluorescent staining of cells pre-stained with Rabbit IgG primary antibodies. It reacts with rabbit IgG and the light chains of other rabbit immunoglobulins. BV421 Anti-Rabbit IgG has minimal crossreactivity with human, mouse, or rat serum proteins. As a second step staining reagent, it is reactive with rabbit polyclonal and monoclonal antibodies.
The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. BD Horizon BV421 has an Ex Max at 407nm and Em Max at 421 nm. The use of a mounting reagent (eg, ProLong™ Gold) is highly recommended to maximize the photostability of BV421.
Specifications
IgG | |
Polyclonal | |
Brilliant Violet 421 | |
Goat | |
0.1 mg | |
Secondary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry, Immunofluorescence | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Rabbit | |
Ig |