missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
338.00€ - 484.87€
Specifications
Antigen | GMPS |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18242371
|
Bio-Techne
NBP2-56670 |
100 μL |
484.87€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18670447
|
Novus Biologicals
NBP2-56670-25ul |
25 μL |
338.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GMPS Polyclonal specifically detects GMPS in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GMPS | |
Polyclonal | |
Rabbit | |
metabolism | |
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein | |
GMPS | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
8833 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AGGDLKDGHHHYEGAVVILDAGAQYGKVIDRRVRELFVQSEIFPLETPAFAIKEQGFRAIIISGGPNSVYAEDAPWFDPAIFTIGKPVLGICYGMQMMNKVFGGTVHKKSVREDGVFNISVDNTCSLFRGLQKEEVVLLT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only