missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547154
This item is not returnable.
View return policy
Description
In Western blots, approximately 25% cross-reactivity with recombinant rat GM-CSF is observed, approximately 5% cross-reactivity with recombinant feline GM-CSF and recombinant porcine GM-CSF is observed, and less than 1% cross-reactivity with recombinant human GM-CSF and recombinant mouse GM-CSF is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
GM-CSF Polyclonal antibody specifically detects GM-CSF in Canine samples. It is validated for Immunocytochemistry, Neutralization, Western Blot.Specifications
GM-CSF | |
Polyclonal | |
Unconjugated | |
CSF2 | |
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim | |
Goat | |
Antigen affinity chromatography | |
RUO | |
403923 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P48749 | |
CSF2 | |
E. coli-derived recombinant canine GM-CSF Ala18-Lys144 | |
100 μg | |
Primary | |
Canine | |
Antibody | |
IgG |