missing translation for 'onlineSavingsMsg'
Learn More

Glycidyl 4-methoxyphenyl ether, 98%, Thermo Scientific Chemicals

Product Code. 11438647
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2211-94-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00005135
InChI Key AVWGFHZLPMLKBL-UHFFFAOYNA-N
Synonym glycidyl 4-methoxyphenyl ether, 2-4-methoxyphenoxy methyl oxirane, 2,3-epoxypropyl-4-methoxyphenyl ether, p-methoxyphenyl glycidyl ether, methoxyphenyl glycidyl ether, glycidyl p-methoxyphenyl ether, 2-p-methoxyphenoxymethyl oxirane, 4-methoxyphenoxy methyl oxirane, anisole, p-2,3-epoxypropoxy
PubChem CID 16646
IUPAC Name 2-[(4-methoxyphenoxy)methyl]oxirane
SMILES COC1=CC=C(OCC2CO2)C=C1

Specifications

Melting Point 45°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Glycidyl 4-methoxyphenyl ether

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'