missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Solution, Honeywell

Product Code. 15641480
Click to view available options
Quantity:
1 L
5 L
25 L
70 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.252g/mL at 25°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Linear Formula HOCH2CH(OH)CH2OH · aq
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Formula Weight 92.09g/mol (anhydrous basis)
Percent Purity 84 to 88%
Grade puriss.
Chemical Name or Material Glycerol Solution, Meets analytical specification of Ph. Eur. BP
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'