missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glucose monohydrate, Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Wide selection of support materials and multiple cartridge/column sizes
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- B&J’s reliable quality
Specifications
Specifications
Packaging | Plastic Bottle |
Quantity | 6 x 1 kg |
Residual Solvents | Complying |
Percent Purity | 97.5 to 102.0% |
Grade | Reagent Pharmacopeia Organics |
Physical Form | Powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'