missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Glove Box Gloves, Sleeves and Hands

Product Code. 17897097 Shop All Labconco Corporation Products
Click to view available options
Material:
Butyl
Neoprene
Size:
9
9.75
19
Color:
Black
Thickness (English):
15 mil
17 mil
This item is not returnable. View return policy
This item is not returnable. View return policy

Glove box gloves, sleeves and hands are available in a variety of sizes, materials and thicknesses to suit your specific application. Choose from neoprene, latex, CSM or butyl.

Glove Box Gloves - One pair/pack, one-piece, seamless, with O-rings

  • Neoprene - Size 9.75
    • 17 mil thickness
  • CSM (Chlorosulfonated polyethylene rubber) - Available in sizes 8.5 or 9.75
    • Also known as Hypalon
    • Resistant to abrasion, ozone and oxidizing chemicals
    • 15 mil thickness
  • Butyl - Available in sizes 8.5 and 9.75
    • Provides high impermeability to water vapor, gases and toxic chemicals
    • Resistant to oxygenated solvents and most oxidizing chemicals
    • 15 mil thickness

Glove Box Sleeves

  • Neoprene Straight Sleeves
    • For use with Neoprene or Latex Hands
    • With O-rings and springs

Glove Box Hands - For use with neoprene straight or accordian sleeves

  • Neoprene available in size 7.5, 9 or 10
    • 17 mil thickness
    • Lightweight and embossed with firm hold finish
  • Latex available in size 7 or 9
    • 9 mil thickness
    • White, surgical-type

Specifications

Color Black
Hand Ambidextrous
Material Neoprene
Size 19
Sterility Non-sterile
Manufacturing/QMS Audit Standards ISO 9001
Model 5005800
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.