missing translation for 'onlineSavingsMsg'
Learn More

Gliclazide, Thermo Scientific Chemicals

Product Code. 15567719
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21187-98-4
Molecular Formula C15H21N3O3S
Molecular Weight (g/mol) 323.41
MDL Number MFCD00409893
InChI Key BOVGTQGAOIONJV-UHFFFAOYNA-N
Synonym gliclazide, glimicron, diamicron, nordialex, diaikron, gliclazida, gliklazid, glyclazide, gliclazidum inn-latin, gliclazida inn-spanish
PubChem CID 3475
ChEBI CHEBI:31654
IUPAC Name 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-(4-methylphenyl)sulfonylurea
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=O)NN1CC2CCCC2C1

Specifications

Melting Point 163.0°C to 169.0°C
Loss on Drying 0.5% max. (1 g, 105°C)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf, slightly soluble in methanol, ethanol
Formula Weight 323.41
Percent Purity ≥98.5% (HPLC)
Chemical Name or Material Gliclazide
compliance-icons
Product Identifier
  • Gliclazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'