Learn More
Lenz Laborglasintrumente™ Glass Stopcock Plug
Click to view available options
Plug Bore:
2.5 mm
4.0 mm
Connectors:
Plug NS 12.5
Plug NS 14.5
Plug NS 18.8
Unit Size:
Each
Description
- For stopcocks
- Straight bore
- Glass
Specifications
Specifications
| Plug Bore | 4.0 mm |
| Material | SBW |
| No. per Pack | 1 |
| Connectors | Plug NS 14.5 |