Learn More
Heidolph instruments Glass Sheath for Sensor Temperature PT1000
Accessories for Heidolph MR Series Hei-Tec Hotplate Stirrers that are ideal in research and scientific laboratory environments.
Brand:  Heidolph instruments 509-67920-00
Product Code. 10639922
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  •  Accessories for Heidolph MR Series Hei-Tec Hotplate Stirrers 
  • Pt 1000 Temperature Sensor is glass coated for improved chemical resistance
Specifications
Specifications
6 x 250 mm