Learn More
Cytiva Glass Plate
Glass plate for use with the miniVE Vertical Electrophoresis System.
Brand: Cytiva SE262P-5
111.83 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Description
- Glass
- 10 x 10.5 cm
- miniVE Vertical Electrophoresis System Accessory
Specifications
Glass | |
10 x 10.5 cm |