Learn More
BUCHI Glass Hood
Use this hood with the Wet Digestor B-440. BUCHI Glass Hood as part of the Wet Digestor B-440 is designed for parallel pre-incineration of samples.
Brand:  BUCHI 036081
Product Code. 10037031
  • 1415.00€ / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Description
  • Glass hood for the Wet Digester B-440
  • Hot fumes rise into the glass hood and are safely withdrawn by the suction flow of the Scrubber
Specifications
Specifications
Glass Hood
B-440 Wet Digester