Learn More
DWK Life Sciences Kimble™ Glass-filled Nylon Connectors with PTFE Seals
Kimble™ Glass-filled Nylon Connectors with PTFE Seals are threaded connectors used for coupling two items with a GPI finish.
Brand:  DWK Life Sciences 747205-1820
 View more versions of this product
Product Code. 11892933
  • 42.60€ / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Connectors have PTFE seals and FKM O-Rings
  • Blue glass-filled nylon
  • For use to 150°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
To connect standard rotary evaporator traps to vials
18-400 to 20-400
Connector
PTFE seals
Nylon