Learn More
Rogo Sampaic™ Glass Coplin Staining Jar
Holds 10 microscope slides of approx. 76 x 26mm. Rogo Sampaic™ Glass Coplin Staining Jars are manufactured from soda lime glass.
Brand:  Rogo Sampaic™ BPL005
Product Code. 11702654
  • 38.00€ / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Clear glass
  • High wall shape
  • With screw cap
Specifications
Specifications
Glass
5
45 mm
105 mm
Coplin Staining Jar
45 x 45 mm
45 mm