Learn More
Gilson™ MACROMAN™ Pipette Controller Accessory, Pipette Mount
Efficient solution for pipetting up to 100mL at once
Brand: Gilson™ F1077505
22.33 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- Pipette mount is used with MACROMAN™ Pipette Controller
Specifications
Pipette Mount | |
Macroman pipette controller | |
1/Cs. | |
1 Year |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
New Offer Save Up to 92%
Product Content Correction
Product Title
Gilson™ MACROMAN™ Pipette Controller Accessory, Pipette Mount > Pipette mount