Learn More
Gilson™ PIPETMANTM TIPACKTM Racked Pipet Tips
GREENER_CHOICE
Product Code. 12679581 Shop All Gilson Products
Click to view available options
Length (Metric):
14.5 cm
32.2 cm
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
100 to 1200 μL
2 to 200 μL
20 to 300 μL
500 to 5000 μL
Model:
D10
D1000
D1200
D200
D300
D5000
DL10
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 600
Pack of 960
Product Code. 12679581
Brand: Gilson™ F171101
Only null left
Add to basket
Edge
Add to basket
Engineered to be functional, convenient and robust. GilsonTM PIPETMANTM TIPACKTM Racked Pipet Tips are available in sterilized or autoclavable packages.
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Requires low tip-mounting force.
  • Robust and designed to withstand autoclaving.
  • Sterility assurance level SAL 10-6 guaranteed, using gamma irradiation.
  • All packs are wrapped in medical device quality plastic which provides a complete shield against contamination.
  • Indicator dots used on Gilson products undergo an irreversible color change upon sterilization; yellow gamma dots turn red when products have been properly exposed.
  • TIPACKs are securely sealed with a sticker; a full product description is given on the TIPACK for traceability and easy identification.
  • Available in sterilized or autoclavable packages.
  • Manufactured in a clean room and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Easy identification with color-coded packaging to meet GLP requirements.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Model D10
Length (English) 5.71 in.
Length (Metric) 14.5 cm
Clean Claims DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
Format Rack
Tip Style Standard
Quantity 960 (10x96)
Product Type Pipet Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Pipetman Tipack
Material Polypropylene
Graduations 2 μL, 10 μL