Learn More
Gilson™ PIPETMAN™ F Fixed Volume Pipette
GREENER_CHOICE
Product Code. 10676073 Shop All Gilson Products
Click to view available options
Volume (Metric):
2 μL
5 μL
10 μL
20 μL
25 μL
50 μL
100 μL
200 μL
250 μL
300 μL
400 μL
500 μL
1000 μL
5000 μL
Accuracy:
±0.08μL
±0.1μL
±0.2μL
±0.25μL
±0.4μL
±0.8μL
±1.6μL
±3μL
±3.5μL
±3.6μL
±30 μL
±4μL
±8μL
Model:
F10
F100
F1000
F2
F20
F200
F25
F250
F300
F400
F5
F50
F500
F5000
Unit Size:
Each
Product Code. 10676073
Brand: Gilson™ F123606
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
The PIPETMAN™ F Fixed Volume Pipette is a dedicated fixed-volume air-displacement pipet that guarantees no risk of volume-selection errors.
  • Dedicated fixed-volume pipette assures reliable and consistent results.
  • Volume can be adjusted by users in the laboratory for dense or viscous fluids.
  • Single-volume mechanism provides a low-cost alternative for clinical diagnostics, quality control and routine testing.
  • Gilson maximum permissible errors are guaranteed only when PIPETMAN pipettes are used with the recommended Gilson PIPETMAN Tips.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy ±8μL
Certifications/Compliance GLP
Compatible Tips D1000, DF1000ST, DF1200ST tip
Autoclavable Autoclavable
Color Blue
Disposable Reusable
For Use With D1000, DF1000ST, DF1200ST Tip
Material Polypropylene
Model F1000
Number of Channels 1
Sterility Non-sterile
Type Fixed Volume Pipette
Volume (Metric) 1000 μL
For Use With (Equipment) D1000, DF1000ST, DF1200ST tip
Includes Stainless Steel piston and PVDF handle
Warranty 3 Years
Length (English) 4.25 in.
Length (Metric) 10.8 cm
Width (English) 12.48 in.
Width (Metric) 31.7 cm
Height (English) 1.42 in.
Height (Metric) 3.6 cm
Fit for Purpose Certified Quality