Learn More
Gilson™ MICROMAN™ Positive-Displacement Pipets
Product Code. 10002082 Shop All Gilson Products
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
100 to 1000 μL
20 to 50 μL
3 to 25 μL
50 to 250 μL
Model:
M10
M100
M1000
M25
M250
M50
Compatible Tips:
C10
C100
C1000
C25
C250
C50
Unit Size:
Each
Product Code. 10002082
Brand: Gilson™ F148501
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Ensures accuracy and precision when pipetting problem liquids, and offers full protection against cross contamination. Gilson™ MICROMAN™ Positive-Displacement Pipets are 3 to 4 times better than an air displacement pipet when pipetting viscous or volatile liquids.
  • Piston is located inside the tip or capillary preventing pipet contamination
  • Color-coded push-button allows for easy identification of different models
  • Dual function push-button that also ejects capillary pistons
  • Comfortable PVDF handle resistant to aggressive chemicals
  • Accurately measure high vapor pressure liquids without leakage or degradation of the tip
  • M10 and M100 models are fully autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips C10
Autoclavable Autoclavable
Model M10
Number of Channels 1
Type Positive Displacement Pipette
Volume (Metric) 1 to 10 μL
For Use With (Application) Pipetting of viscous and dense liquids