Learn More
Gilson™ Accessory for PIPETMANTM Complete Pipetting System- SingleTM Pipet Holder
Shop All Gilson Products
Click to view available options
:
Single Pipet Holder; Sticks to any smooth, dry surface without damage
Description
- Sticks to any smooth, dry surface without damage
- Resistant to chemicals and UV light
- Holds any Gilson Pipette except the PIPETMANTM Concept
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Specifications
Content And Storage | 1x Single pipette holder |
Color | White |
For Use With | Pipettes |
Material | Polypropylene |
Type | Pipette Holder |
For Use With (Equipment) | Pipetteman™ complete pipetting system |
Includes | 1x Single pipette holder |
Packaging Type | Box |
Quantity | 1 |
Dimensions (L x W x H) | 50 X 95 X 50 mm |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Promotions
-
Multiple Manufacturers
Focus on Agri-Science Testing
Use promotion code 24136 to get amazing savings.Promo Code: 24136Expires: 2025-10-26
Days
49Hours
19Minutes
8Seconds
32 -
Multiple Manufacturers
Fisherbrand Sweet Deals on Top Products
Get up to 60% off select Fisherbrand productsPromo Code: 24168Expires: 2025-09-30
Days
23Hours
19Minutes
8Seconds
32 -
Multiple Manufacturers
New Offer Save Now
Product Content Correction
Product Title
Gilson™ Accessory for PIPETMANTM Complete Pipetting System- SingleTM Pipet Holder >