Learn More
Brady™ GHS Symbol - Health Hazard, Individual Markers on a Roll
Pre-printed labels to identify your chemical product containers.
Brand:  Brady™ 834182
Product Code. 15462044
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • High-quality labels that can withstand the most demanding environments
  • Highly resistant to bad weather, abrasion and chemical products
Specifications
Specifications
Gloss
250/Pk.
21 mm
21 mm
Diamond
Polyester
Permanent Acrylic
0.099 mm
-40°C to 110°C
Black on White