missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Gentamicin sulfate

Product Code. 15585136 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Product Code. 15585136

Brand: Thermo Scientific Chemicals 455310250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Gentamicin sulfate, CAS # 1405-41-0, is an aminoglycoside mainly used to inhibit the growth of Gram-negative bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Gentamicin sulfate is a bactericidal aminoglycoside antibiotic which shows activity against Gram-negative bacteria
  • It works by inhibiting protein biosynthesis through binding the bacterial ribosome 30S subunit

Applications

  • Gentamicin sulfate has been used in studies to determine wound healing due to its antibiotic and antifungal properties
  • It has laboratory research applications in the study of a number of clinical conditions, including actinomycotic infection, appendicitis, cystic fibrosis, osteomyelitis, pelvic inflammatory disease, plague, gonorrhea, and tularemia
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-41-0
Synonym Garamycin, Gentamicin Sulfate

Specifications

Type Gentamicin Sulfate
Melting Point 218.0°C to 237.0°C
Infrared Spectrum Authentic
Water 15% max.
Quantity 25 g
Solubility Information Solubility in water: soluble.
Percent Purity ≥590 IU/mg (Gentamycin)
Chemical Name or Material Gentamicin sulfate
compliance-icons
Product Identifier
  • Gentamicin sulfate
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.