Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer Accessories
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
Brand: Thermo Scientific™ 840-278000
664.38 EUR valid until 2025-03-30
Use promo code "24134" to get your promotional price.
This item is not returnable.
View return policy
Description
Get all of the tools you need to facilitate your analytical process with accessories for the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer.
- Printer attaches directly to instrument behind screen
- Easy drop-in paper re-loading
- Uses 80mm wide thermal paper rolls
Specifications
This snap-on thermal printer attaches easily anddirectly to the back of the instrument and uses 80mm wide thermal drop in paper rolls. It can also be used with regular or self stick printer paper. | |
Direct hard-copy printing of results from the instrument onboard software control. | |
1 roll thermal paper |
Thermal Snap-on Printer | |
GENESYS 30 Visible Spectrophotometer |