Learn More
Applied Biosystems™ GeneScan™ Installation Standard DS-33 , [6FAM™, VIC™, NED™, PET™ dyes]
Using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347.
Brand:  Applied Biosystems™ 4330397
Product Code. 16300824
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Includes: 4 Tubes of fluorescently labeled pooled PCR products, 1 tube of GeneScan -500 LIZ Size Standard, and 1 tube of loading buffer
Description
Description
The DS-33 GeneScan™ Installation Standards using 6FAM™, VIC™, NED™ and PET™ dyes contain pooled PCR products amplified from microsatellite loci present in CEPH individual 1347. The kit also contains a tube of GeneScan™ 500 LIZ™ Size Standard.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Specifications
This kit contains 4 tubes of fluorescently labeled pooled PCR products and 1 tube of GeneScan 500 LIZ Size Standard.
Store the kit at -15 to -25°C until ready to use. Upon thawing, store the GeneScan 500 LIZ Size Standard tube at 2–8°C.
FAM, NED PET, VIC
FAM, NED, PET, VIC
Dry Ice
3730xl DNA Analyzer, 3730 DNA Analyzer
Installation Standard
1 kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.