Learn More
SI Analytics™ TZ 1752 Generating Electrode
SI Analytics™ TZ 1752 Generating Electrode does not come equipt with a diaphragm junction.
Brand:  SI Analytics™ 285216660
Product Code. 12655706
  • 867.00€ / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Electrode
  • For use with TitroLine™ KF and TitroLine™ KF Trace
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Generating Electrode
TitroLine KF and TitroLine KF trace
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers