Learn More
SI Analytics™ TZ 1753 Generating Electrode
SI Analytics™ TZ 1753 Generating Electrode is equipt with a diaphragm junction.
Brand:  SI Analytics™ 285216650
Product Code. 10488104
Description
Description
  • Electrode
  • For use with TitroLine™ KF and TitroLine™ KF Trace
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Generating Electrode
TitroLine KF and TitroLine KF trace