Learn More
Thermo Scientific™ GeneBLAzer™ ENDRB-NFAT-bla HEK 293T Cells
Product Code. 10542764
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
Product Code. 10542764
Brand: Thermo Scientific™ K1727
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells contain the human Endothelin Type B Recpetor (EDNRB) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line.
GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells contain the human Endothelin Type B Recpetor (EDNRB) stably integrated into the CellSensor™ NFAT-bla HEK293T cell line. CellSensor™ NFAT-bla HEK293T cells (Cat. No. K1538) contain a beta-lactamase reporter gene under control of the Nuclear Factor of Activated T-cells (NFAT) response element.
The GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells are functionally validated for Z' and EC50 concentrations of ET-1. In addition, GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
The ENDRB receptor mediates signaling of endothelin proteins that regulate several critical biological processes, including the development and function of blood vessels, the production of certain hormones, and the stimulation of cell growth and division. Defects in EDNRB mediated signaling have been implicated in a wide range of diseases, including cancer, hypertension and cardiovascular disorders.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_000115.1
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells
• GeneBLAzer™ EDNRB-NFAT-bla HEK293T Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ GeneBLAzer™ EDNRB-NFAT-bla HEK293T cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol EDNRB
Includes EDNRB-NFAT-bla HEK293T cells, EDNRB-NFAT-bla HEK293T Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry EDNRB
Type Cell Line
Cell Line 293T (HEK)
System Type GeneBLAzer™
Druggable Target GPCR