Learn More
Thermo Scientific™ GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
Product Code. 10389054
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
Product Code. 10389054
Brand: Thermo Scientific™ K1837
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line.
GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells contain the human Adenosine A3 Receptor (ADORA3) stably integrated into the GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cell line. The GeneBLAzer™ Ga15-NFAT-bla CHO-K1 cells (Cat. No. K1537) contain a beta-lactamase reporter gene under control of the NFAT Response Element and a promiscuous G protein, Ga15. GeneBLAzer™ ADORA3-Ga15-NFAT-bla CHO-K1 cells are functionally validated for Z'-factor and EC50 concentrations of NECA.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_000677.2
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells
• GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 Assay Protocol
• Certificate of Analysis
The GeneBLAzer™ ADORA3-Gα15-NFAT-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol ADORA3
Human Protein Class G-Protein Coupled Receptor (GPCR)
Includes ADORA3-Gα15-NFAT-bla CHO-K1 cells, ADORA3-Gα15-NFAT-bla CHO-K1 Assay Protocol, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 8 x 106 Cells
Readout End Point
Target Entry ADORA3
Type Dividing Cells
Cell Line CHO-K1
System Type Tango™
Druggable Target GPCR