Learn More
Thermo Scientific™ GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 Cells
Product Code. 10626414
Click to view available options
Quantity:
2 x 106 Cells
Unit Size:
Each
Product Code. 10626414
Brand: Thermo Scientific™ K1530
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells contain the human Adenosine Receptor A2A (ADORA2A) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line.
GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells contain the human Adenosine Receptor A2A (ADORA2A) stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. No. K1535) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE) response element.
The GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells are functionally validated for Z' and EC50 concentrations of NECA. In addition, GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
ADORA2A has been shown to have many roles throughout the body. ADORA2A is involved in pain and inflammation regulation, inhibition of platelet aggregation, blood pressure regulation and contributes to ischemic brain damage. Selective antagonists of the ADORA2A pathway are being tested as adjuvants to dopaminergic drugs to treat Parkinson's disease and schizophrenia.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number NM_000675
Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells
• GeneBLAzer™ ADORA2A-CRE-bla CHO-K1
• Certificate of Analysis
The GeneBLAzer™ ADORA2A-CRE-bla CHO-K1 cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol ADORA2A
Includes ADORA2A-CRE-bla CHO-K1 cells, ADORA2A-CRE-bla CHO-K1, Certificate of Analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer
Quantity 2 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Target Entry ADORA2A
Type Dividing Cells
Cell Line CHO-K1
System Type GeneBLAzer™
Druggable Target GPCR