missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneAmp™ 10X PCR Gold Buffer & MgCl2

Applied Biosystems GeneAmp 10X PCR Gold Buffer is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification.

165.00€ - 378.00€

Specifications

Format Kit
Polymerase AmpliTaq Gold DNA Polymerase
Product Type 10X PCR Gold Buffer
For Use With (Application) Standard PCR
Concentration 10X
View More Specs

Products 2
Product Code Brand Volume Price Quantity & Availability  
Product Code Brand Volume Price Quantity & Availability  
10536745
View Documents Promotion Details
Applied Biosystems™
4379874
2 x 75 mL
378.00€
150mL
Please to purchase this item. Need a web account? Register with us today!
10608095
View Documents Promotion Details
Applied Biosystems™
4306898
6 x 1.5 mL
165.00€
9mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applied Biosystems GeneAmp 10X PCR Gold Buffer and MgCl2 is formulated to provide flexible, efficient activation of AmpliTaq Gold DNA Polymerase, resulting in highly specific, robust PCR amplification. The ionic strength and pH of GeneAmp 10X PCR Gold Buffer has been optimized to provide a wider activation temperature and time range when used in with AmpliTaq Gold DNA Polymerase.

When GeneAmp 10X PCR Gold Buffer and the MgCl2 solution are used together, there is greater versatility in optimizing magnesium ion concentrations for optimal PCR amplification with any specific set of primers and DNA template.

Features

  • Magnesium ion–free
  • Gelatin-free
  • High versatility in optimizing magnesium ion concentration
  • MgCl2 solution provided separately
Specifications

Specifications

Kit
10X PCR Gold Buffer
10X
AmpliTaq Gold DNA Polymerase
Standard PCR
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.