Learn More
Agilent BioTek Gen5 Data Collection and Analysis Software
Shop All Agilent BioTek Products
Click to view available options
Model:
Gen5 AL2 Data Analysis Software
Gen5 AL3 Data Analysis Software
Gen5 AL4 Data Analysis Software
Gen5 IVD Data Analysis Software
Unit Size:
Each
Description
- Quick launch buttons on the welcome screen provide links to recently used files, pre-programmed protocols and other key functions such as Quick Read and System Test
- StepWise tools allow limitless combinations of procedural steps and data reduction criteria for a wide variety of applications in a logical and clearly defined sequence that is easily viewed on the screen
- Analyze data on a single plate or across multiple plates in an experiment
- Report and PowerExport builders are fast and flexible output tools to produce publication-ready reports using data matrices, statistics tables, protocols and plate information
- For compliance to 21 CFR Part 11, Gen5 Secure provides system, data and protocol protection
- Multiple user definitions and permission levels make it easy to comply with laboratory policies
- Windows XP or Windows 2000 (Professional Editions)
- PentiumIII-Class PC (or compatible) processor (500MHz or higher)
- 512 MB RAM or higher
- 2 GB hard drive space or higher
- XGA resolution at 1024 x 768 or higher
- CD-ROM readable drive
- Keyboard
- Mouse
- Microsoft Internet Explorer v 5.0 or higher (for online help)
- Microsoft Excel 2000 or later (for PowerExport feature)
Specifications
Specifications
| Model | Gen5 AL3 Data Analysis Software |
| For Use With (Equipment) | Imaging Multi-Mode Readers |
| Type | Data Collection and Analysis Software |