Learn More
Epredia™ Gemini™ AS Automated Slide Stainer
Shop All Epredia ProductsDescription
The Gemini AS Automated Slide Stainer delivers high-throughput staining capabilities to the busy histology laboratory. With its new intuitive touch-screen interface, operation is easy-to-learn and simple-to-use.
The unique dual-level design, with integrated power supply and touch-screen, ensures maximum space savings. With its small footprint, the Gemini AS Stainer can be placed near the sectioning area for optimal workflow. The intelligent look-ahead control software provides the ability to run up to 12 slide racks simultaneously with minimal reduction in speed. The “urgent” feature enables a rack to be given priority for MOHS surgery or similar STAT needs. The five heated stations provide maximum throughput without the need for a separate drying oven.
Multidimensional Design
- Two-tier design occupies minimal bench space
- Integrated color touch-screen with intuitive software offers quick learning and simple operation
- Scheduler software allows multiple protocols and slide racks to be run simultaneously at optimal speed
- User easily sees progress of slides and placement of reagents with built-in LED lighting
- Fully editable protocols and stain sets provide flexibility in programming
- Fume extraction, battery backup and lockable slide rack ensure safety for the slides and the operator
- 26 or 31 reagent stations accommodate multiple protocols without changing reagents
- One-touch on-screen view of reagent status provides simple reagent monitoring
Performance
- Battery backup with 40 minutes of continual staining power
- Download protocols and log files via USB connectivity
- Load-on-demand flexibility with STAT capability
- Units available with or without five independently heated stations
- Six running water stations
- Four configurable doors for convenient loading and unloading
- Fume control easily managed with carbon filters or vent adapter kit
- Internal flow regulators and anti-siphon devices to protect water supply
- Lockable baskets to ensure secure slides, even upside down
- Auto-return recalls any loaded basket to the load door
- Passcode protection to secure customized protocols
Specifications
Specifications
| Product Type | Slide Stainer, unheated |
| Slide Capacity | 20 Slides/Basket |
| Dimensions (L x W x H) | 79 x 72 x 88 cm (31 x 28 x 35 in.) |
| Height (English) | 35 in. |
| Height (Metric) | 88 cm |
| Length (English) | 31 in. |
| Length (Metric) | 79 cm |
| Width (English) | 28 in. |
| Width (Metric) | 72 cm |
| Frequency | 50/60 Hz |
| Show More |