Learn More
Fisherbrand™ Gel Casting Tray
Forms leak proof seal against inner walls.
Brand: Fisherbrand™ HU6-UT
This item is not returnable.
View return policy
Description
- Silicone gaskets to line casting tray
- Forms leak proof seal against inner walls of running chamber when casting tray is turned at 90° to direction of electrophoresis
Specifications
Gel Casting Tray | |
Acrylic | |
60 mm |
Leakproof Silicone Gasket | |
75 mm |