Learn More
Velp Scientifica™ GDE Enzymatic Digester
Enzymatic digestion for dietary fiber analysis by enzymes heating. It offers non-stop sample mixing for homogeneity.
Brand:  Velp Scientifica™ SA30400209
Product Code. 12891683
  • 2540.00€ / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The analytical procedure for the determination of dietary fiber involves a series of digestions of the sample by thermostable enzymes using the enzymatic digester GDE.
  • It operates in accordance with the official AOAC method for the determination of total dietary fiber.
  • The magnetic stirrer of GDE ensures homogeneity thanks to a continuous and constant stirring of the sample while avoiding overheating that could compromise the success of enzymatic digestions.
  • Set the timer for unattended operation, you can quickly set the operating time and focus on other tasks.
  • The transparent polycarbonate tank ensures visibility and enhanced resistance up to 105 °C; whilst the use of deionized water eliminates any possible creation of limestone.
  • Outstanding temperature regulation is possible with GDE: through an intuitive digital display it is possibile to set the desired temperature, with an excellent accuracy of only ± 0.2 °C.
Specifications
Specifications
6.2 kg