missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Garcin™ Bactinyl Disinfectant Detergent
Use Garcin™ Bactinyl Disinfectant Detergent as a janitorial cleaner and disinfectant. Its active substances include peroxides, quaternary ammoniums and ethanol.
Brand: Garcin™ LB920670
This item is not returnable.
View return policy
Description
- Sterile
- Rinse not justified if thorough drying
- Odorless
- Inactivation of the HIV virus in 5 minutes
- Approved for food contact
Specifications
Bactinyl Disinfectant Detergent | |
10 ±1 | |
EN 1040, EN 1276, NF T 72-151 spectrum 5, NF T 72-190 NF T 72-171et, NF EN 1275 and NF EN 1650, NF T 72-180, EC 0398 | |
4°C | |
5 L |
Liquid | |
Opalescent | |
Can | |
1 Ea. |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- bctinyl perfumed
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, didecyldimethylammonium chloride, benzalkonium chloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction