Learn More
Garcin™ Bactinyl Disinfectant Detergent
Use Garcin™ Bactinyl Disinfectant Detergent as a janitorial cleaner and disinfectant. Its active substances include peroxides, quaternary ammoniums and ethanol.
Brand:  Garcin™ LB920723
 View more versions of this product
Product Code. 11788743
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Sterile
  • Rinse not justified if thorough drying
  • Odorless
  • Inactivation of the HIV virus in 5 minutes
  • Approved for food contact
Specifications
Specifications
Bactinyl Disinfectant Detergent
10 ±1
EN 1040, EN 1276, NF T 72-151 spectrum 5, NF T 72-190 NF T 72-171et, NF EN 1275 and NF EN 1650, NF T 72-180, EC 0398
4°C
500 mL
Liquid
Opalescent
Bottle
1 Ea.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.