missing translation for 'onlineSavingsMsg'
Learn More

GAPDH Monoclonal Antibody (1E6D9), CoraLite488, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech CL488-60004-100UL

Product Code. 17505786

  • 189.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GAPDH Monoclonal antibody specifically detects GAPDH in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

GAPDH
Monoclonal
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P62736, P62737, P62738
Mouse
100 μL
Primary
Human, Mouse, Rat
Liquid
Flow Cytometry, Western Blot
1E6D9
CoraLite 488
ACTA2
38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; bb02e05; cb350; cb609; CDABP0047; EC 1.2.1.12; epididymis secretory protein Li 278; epididymis secretory sperm binding protein Li 162eP; fb71f08; fk58c09; G3PD; G3PDH; GAPD; GAPD2; gapdh; GAPDH2; GAPDH-2; GAPDHS; Gapds; Gapd-s; glceraldehyde-3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase, testis-specific; glyceraldehyde phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase (G3PDH); glyceraldehyde-3-phosphate dehydrogenase 2; glyceraldehyde-3-phosphate dehydrogenase GAPDH; glyceraldehyde-3-phosphate dehydrogenase like-17 protein; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; glyceraldehyde-phosphate-dehydrogenase; glycerine aldehyde 3-phosphate dehydrogenase; HEL-S-
Protein A
RUO
11475, 59, 81633
-20° C, Avoid Freeze/Thaw Cycles, store in dark
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.