missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Tubulin Antibody (TU-30), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-574AF647
This item is not returnable.
View return policy
Description
gamma Tubulin Monoclonal specifically detects gamma Tubulin in Human, Mouse, Rat, Porcine, Bovine, Chicken, Plant, Protozoa samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| gamma Tubulin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| P23258 | |
| TUBG1 | |
| C-terminal peptide of gamma-tubulin counjugated to KLH. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Centrosome Markers, Cytoskeleton Markers | |
| 7283 | |
| Human, Mouse, Rat, Pig, Bovine, Chicken, Plant, Protozoa | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| TU-30 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| gamma-1-tubulin, Gamma-tubulin complex component 1, GCP-1, TUBG, TUBGCP1, tubulin gamma-1 chain, tubulin, gamma 1, tubulin, gamma polypeptide | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody TU-30 recognizes C-terminal peptide sequence EYHAATRPDYISWGTQ (aa 434-449) of gamma-tubulin, a 48 kDa structural constituent of cytoskeleton and microtubule organizing center (MTOC). | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction