Learn More
WTW™ Galvanic Dissolved Oxygen Sensors (universal)
Get accurate testing results for water and waterous solutions with these membrane-covered galvanic dissolved oxygen sensors that are low maintenance and no polarizarion time required.
Brand:  WTW™ 201547
 View more versions of this product
Product Code. 10235731
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Dissolved oxygen sensor with integrated temperature compensation and membrane leak monitoring
  • Can be used in the lab as well as in the field
  • Features include a quick response time (t99<60 s) and low-maintenance operation
  • Fields of use include surface waters, measurements in wastewater treatment plants, BOD directions
  • Filled with electrolyte and achieves a long dwelling time up to 6 months
  • Calibration vessel is part of the delivery
  • For use with conductivity and multiparameter meters
  • Includes 1.5m L Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 m
10m Length Cable Cleaning solution, abrasive foil, 3 exchange membrane heads and electrolyte
Conductivity and multiparameter meters
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products