Learn More
Thermo Scientific™ Gable Cover for VersaCool™ Refrigerated Circulating Bath
Reversible hinged cover for water, water/glycol applications to be used with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Brand:  Thermo Scientific™ 230000003
Product Code. 15499529
  • 577.88€ / Each
Estimated Shipment: 30-10-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Utilize the gable cover to accommodate larger size beakers for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Optional accessory for the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath. This reversible hinged cover allows for larger size beakers and:
  • Reduces moisture and directs condensate back into bath
  • Ideal for water, water/glycol applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gable Cover-reversible hinged cover for water, water/glycol applications
Thermo Scientific VersaCool Refrigerated Circulating Bath
Gable Cover