Learn More
Furfuryl isocyanate, 97%, Thermo Scientific Chemicals
58.99€ - 193.97€
Chemical Identifiers
CAS | 71189-15-6 |
---|---|
Molecular Formula | C6H5NO2 |
Molecular Weight (g/mol) | 123.11 |
MDL Number | MFCD03180268 |
InChI Key | UIADMYLYGJYUSQ-UHFFFAOYSA-N |
Synonym | 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl |
PubChem CID | 4223811 |
IUPAC Name | 2-(isocyanatomethyl)furan |
SMILES | O=C=NCC1=CC=CO1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15345078
|
Thermo Scientific Acros
456490010 |
1 g |
58.99€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15355078
|
Thermo Scientific Acros
456490050 |
5 g |
193.97€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
71189-15-6 | |
123.11 | |
UIADMYLYGJYUSQ-UHFFFAOYSA-N | |
4223811 | |
O=C=NCC1=CC=CO1 |
C6H5NO2 | |
MFCD03180268 | |
2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl | |
2-(isocyanatomethyl)furan |
Specifications
71189-15-6 | |
1.1480g/mL | |
Authentic | |
Glass Bottle | |
MFCD03180268 | |
2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl | |
O=C=NCC1=CC=CO1 | |
123.11 | |
123.11 | |
Furfuryl isocyanate |
157.0°C to 158.0°C | |
54.5°C | |
96% min. (GC) | |
C6H5NO2 | |
1.148 | |
UIADMYLYGJYUSQ-UHFFFAOYSA-N | |
2-(isocyanatomethyl)furan | |
4223811 | |
97% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k
GHS Signal Word: Danger
- Furfuryl isocyanate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.