Learn More
Löser™ Funnel Cleaning Needle
Funnel cleaning needle for i Osmometer
Brand:  Löser™ 0.01.0005
Product Code. 11940557
  • 1.00€ / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Funnel for cleaning the needle hole of automatic Osmometers
Specifications
Specifications
Funnel Cleaning Needle
Osmometer Type 15/16
Cleaning needle and needle hole
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Löser™ Funnel Cleaning Needle > Product Type: Funnel Cleaning Needle