Learn More
Invitrogen™ Fremanezumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559141
 View more versions of this product
Product Code. 30284575
  • 477.00€ / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Fremanezumab is a biosimilar that targets CALCA .
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fremanezumab Humanized
Recombinant Monoclonal
Unconjugated
fremanezumab-vfrm; LBR-101; PF-04427429; RI-307; RN-307; TEV-48125
Protein A
RUO
Human
Antibody
IgG2SA
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized