Learn More
Euro Protection™ Freedite Low-Cut Safety Shoes
Euro Protection™ Freedite Low-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.
Brand: Euro Protection™ 9FREL40
Description
- Footbridge for torsional rigidity.
- Padding for snug fit.
- Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
- Full grain leather upper, reflective trim at the back.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
- Composite toe cap.
- Non-metallic (composite fabric) puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
Specifications
Black | |
Leather | |
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. | |
Low | |
563 g per shoe in size 8 |
Breathable 3D mesh lining | |
40 | |
Polyurethane, Composite Fabric | |
Composite | |
EN ISO 20345 : 2011 |